Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-O-Acetyl-D-malic Anhydride 97.0+%, TCI America™

Click to view available options
Quantity:
1g
5g
79814-40-7
C6H6O5
158.109
SSWJHSASZZAIAU-SCSAIBSYSA-N
r-+-2-acetoxysuccinic anhydride,r-2-o-acetylmalic anhydride,r-2,5-dioxotetrahydrofuran-3-yl acetate,+-o-acetyl-d-malic anhydride,2,5-furandione, 3-acetyloxy dihydro-, r,o-acetyl d-malic anhydride,s-acetoxysuccinic anhydride,o-acetyl-d-malic anhydride,ksc496o4p,r-acetoxysuccinic anhydride
9815264
[(3R)-2,5-dioxooxolan-3-yl] acetate
CC(=O)OC1CC(=O)OC1=O
79814-40-7
C6H6O5
158.109
SSWJHSASZZAIAU-SCSAIBSYSA-N
r-+-2-acetoxysuccinic anhydride,r-2-o-acetylmalic anhydride,r-2,5-dioxotetrahydrofuran-3-yl acetate,+-o-acetyl-d-malic anhydride,2,5-furandione, 3-acetyloxy dihydro-, r,o-acetyl d-malic anhydride,s-acetoxysuccinic anhydride,o-acetyl-d-malic anhydride,ksc496o4p,r-acetoxysuccinic anhydride
9815264
[(3R)-2,5-dioxooxolan-3-yl] acetate
CC(=O)OC1CC(=O)OC1=O
Quantity 1g
Boiling Point 162°C
Color White
Melting Point 57°C
Formula Weight 158.11
Physical Form Crystal-Powder at 20°C
Percent Purity ≥97.0% (T)
Chemical Name or Material (+)-O-Acetyl-D-malic Anhydride