Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Acetyl-L-carnitine hydrochloride, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
5080-50-2
C9H19ClNO4+
240.704
MFCD00082230
JATPLOXBFFRHDN-QRPNPIFTSA-O
Levacecarnine hydrochloride
123133819
[(2S)-2-acetyloxy-3-carboxypropyl]-trimethylazanium;hydrochloride
CC(=O)OC(CC(=O)O)C[N+](C)(C)C.Cl
5080-50-2
C9H19ClNO4+
240.704
MFCD00082230
JATPLOXBFFRHDN-QRPNPIFTSA-O
Levacecarnine hydrochloride
123133819
[(2S)-2-acetyloxy-3-carboxypropyl]-trimethylazanium;hydrochloride
CC(=O)OC(CC(=O)O)C[N+](C)(C)C.Cl
Infrared Spectrum Authentic
Quantity 5g
Packaging Glass bottle
Color White
Specific Rotation -27° to -31° (20°C, 589nm) (c=1, H2O)
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Merck Index 15, 79
Formula Weight 239.7
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material o-Acetyl-L-carnitine hydrochloride, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

HYGROSCOPIC