Learn More
Medchemexpress LLC O-acetyl-N-((9H-fluoren-9-ylmethoxy)carbonyl)-L-serine | 171778-17-9 | MFCD00077070 | 97.4% | 369.37 g/mol | C20H19NO6 | 100 G

Supplier: Medchemexpress LLC HYW048674100G
Fmoc-Ser(Ac)-OH (Fmoc-O-acetyl-L-serine) is an Fmoc-protected O-acetyl L-serine derivative used as a building block in peptide synthesis and small-molecule research. It is documented for use in the preparation of broad-spectrum coronavirus membrane fusion inhibitors.
- O-acetyl-N-((9H-fluoren-9-ylmethoxy)carbonyl)-L-serine chemical class.
- CAS number 171778-17-9.
- Molecular formula C20H19NO6, molecular weight ~369.37 g/mol.
- Stated purity approximately 97.4% (HPLC).
- Supplied as laboratory research reagent in common pack sizes: 5 g, 10 g, 25 g, 100 g.
- Typical applications: peptide synthesis and amino acid derivative for research use.
- Supporting documents available: product data sheet, certificate of analysis, safety data sheet.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.