Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Anisaldehyde, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
135-02-4
C8H8O2
136.15
MFCD00003308
PKZJLOCLABXVMC-UHFFFAOYSA-N
o-anisaldehyde,benzaldehyde, 2-methoxy,2-anisaldehyde,o-methoxybenzaldehyde,6-methoxybenzaldehyde,salicylaldehyde methyl ether,2-methoxybenzenecarboxaldehyde,benzaldehyde, o-methoxy,2-methoxyphenylformaldehyde,formylanisole, o
8658
2-methoxybenzaldehyde
COC1=CC=CC=C1C=O
135-02-4
C8H8O2
136.15
MFCD00003308
PKZJLOCLABXVMC-UHFFFAOYSA-N
o-anisaldehyde,benzaldehyde, 2-methoxy,2-anisaldehyde,o-methoxybenzaldehyde,6-methoxybenzaldehyde,salicylaldehyde methyl ether,2-methoxybenzenecarboxaldehyde,benzaldehyde, o-methoxy,2-methoxyphenylformaldehyde,formylanisole, o
8658
2-methoxybenzaldehyde
COC1=CC=CC=C1C=O
Boiling Point 238.0°C
Flash Point 117°C
Refractive Index 1.5608
Packaging Glass bottle
Quantity 500g
Infrared Spectrum Authentic
Melting Point 34.0°C to 40.0°C
Color Brown to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3OC6H4CHO
Beilstein 08, IV, 180
Merck Index 10, 5866
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol
Formula Weight 136.15
Physical Form Low Melting Crystalline Mass
Percent Purity 98%
Chemical Name or Material o-Anisaldehyde, 98%
Show More Show Less

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep comfortable for breathing.

Warning

EINECSNumber : 205-171-7

RTECSNumber : BZ2610000

TSCA : TSCA