Alfa Aesar™ o-Benzoic sulfimide, 98+%

Manufacturer: Alfa Aesar™ B2393822

Quantity 100g

 View more versions of this product

Catalog No. AAB2393822

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 81-07-2
    Molecular Formula C7H5NO3S
    Formula Weight 183.18
    MDL Number MFCD00005866
    Beilstein 6888
    Merck Index 14,8311
    Synonym Saccharin; 1,2-Benzothiazol-3(2H)-one 1,1-dioxide
    Chemical Name or Material o-Benzoic sulfimide

    Specifications

    Formula Weight 183.18
    TSCA TSCA
    Molecular Formula C7H5NO3S
    MDL Number MFCD00005866
    CAS 81-07-2
    Beilstein 6888
    Merck Index 14,8311
    Synonym Saccharin; 1,2-Benzothiazol-3(2H)-one 1,1-dioxide
    Melting Point 227° to 230°C
    Quantity 100g
    RTECS Number DE4200000
    Chemical Name or Material o-Benzoic sulfimide
    Assay ≥98%

    Description

    Applications
    o-Benzoic sulfimide is used as a pharmaceutic flavor aid. It is used in high performance liquid chromatographic method for the simultaneous separation and determination of acesulfame potassium, saccharine and aspartame.

    Solubility
    Soluble in water (3.3 g/L at 20°C).

    Notes
    Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit