Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ O-Benzyl-D-tyrosine, 95%

Click to view available options
Quantity:
1g
5g
25g
65733-15-5
C16H17NO3
271.316
MFCD00077104
KAFHLONDOVSENM-OAHLLOKOSA-N
h-d-tyr bzl-oh,r-2-amino-3-4-benzyloxy phenyl propanoic acid,o-benzyl-d-tyrosine,o-benzyl-d-tyr,d-tyrosine bzl-oh,2r-2-amino-3-4-benzyloxy phenyl propanoic acid,h-d-tyr bzl,pubchem18635,r-2-amino-3-4-benzyloxy phenyl propanoicacid,2r-2-amino-3-4-phenylmethoxyphenyl propanoic acid
725704
(2R)-2-amino-3-(4-phenylmethoxyphenyl)propanoic acid
C1=CC=C(C=C1)COC2=CC=C(C=C2)CC(C(=O)O)N
65733-15-5
C16H17NO3
271.316
MFCD00077104
KAFHLONDOVSENM-OAHLLOKOSA-N
h-d-tyr bzl-oh,r-2-amino-3-4-benzyloxy phenyl propanoic acid,o-benzyl-d-tyrosine,o-benzyl-d-tyr,d-tyrosine bzl-oh,2r-2-amino-3-4-benzyloxy phenyl propanoic acid,h-d-tyr bzl,pubchem18635,r-2-amino-3-4-benzyloxy phenyl propanoicacid,2r-2-amino-3-4-phenylmethoxyphenyl propanoic acid
725704
(2R)-2-amino-3-(4-phenylmethoxyphenyl)propanoic acid
C1=CC=C(C=C1)COC2=CC=C(C=C2)CC(C(=O)O)N
Beilstein 5819231
Formula Weight 271.32
Physical Form Solid
Percent Purity 95%
Quantity 1g
Chemical Name or Material O-Benzyl-D-tyrosine

storageNote1 : 2° to 8°C