Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ O-Benzyl-L-serine, 99%

Click to view available options
Quantity:
1g
5g
25g
4726-96-9
C10H13NO3
195.218
MFCD00065937
IDGQXGPQOGUGIX-VIFPVBQESA-N
o-benzyl-l-serine,h-ser bzl-oh,s-2-amino-3-benzyloxy propanoic acid,l-serine, o-phenylmethyl,2s-2-amino-3-benzyloxy propanoic acid,o-phenylmethyl-l-serine,serine, o-phenylmethyl,benzylserine,o-benzylserine #,z-o-benzyl-l-serine
78457
(2S)-2-amino-3-phenylmethoxypropanoic acid
C1=CC=C(C=C1)COCC(C(=O)O)N
4726-96-9
C10H13NO3
195.218
MFCD00065937
IDGQXGPQOGUGIX-VIFPVBQESA-N
o-benzyl-l-serine,h-ser bzl-oh,s-2-amino-3-benzyloxy propanoic acid,l-serine, o-phenylmethyl,2s-2-amino-3-benzyloxy propanoic acid,o-phenylmethyl-l-serine,serine, o-phenylmethyl,benzylserine,o-benzylserine #,z-o-benzyl-l-serine
78457
(2S)-2-amino-3-phenylmethoxypropanoic acid
C1=CC=C(C=C1)COCC(C(=O)O)N
Beilstein 2114846
Formula Weight 195.22
Percent Purity 99%
Melting Point 213°C (decomposition)
Quantity 1g
Optical Rotation +7° (c=1 in 1N HCl)
Chemical Name or Material O-Benzyl-L-serine