Alfa Aesar™ O-Benzyl-L-serine, 99%

Manufacturer: Alfa Aesar™ B2239306

Quantity 5g

 View more versions of this product

Catalog No. AAB2239306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4726-96-9
    Molecular Formula C10H13NO3
    Formula Weight 195.22
    MDL Number MFCD00065937
    Beilstein 2114846
    Synonym (S)-2-Amino-3-(benzyloxy)propionic acid; H-Ser(Bzl)-OH
    Chemical Name or Material O-Benzyl-L-serine

    Specifications

    Formula Weight 195.22
    TSCA TSCA
    Molecular Formula C10H13NO3
    MDL Number MFCD00065937
    CAS 4726-96-9
    Beilstein 2114846
    Melting Point 213°C (dec.)
    Quantity 5g
    Optical Rotation +7° (c=1 in 1N HCl)
    Synonym (S)-2-Amino-3-(benzyloxy)propionic acid; H-Ser(Bzl)-OH
    Chemical Name or Material O-Benzyl-L-serine
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit