Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-Benzyl-L-serine, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
4726-96-9
C10H13NO3
195.22
IDGQXGPQOGUGIX-VIFPVBQESA-N
o-benzyl-l-serine,h-ser bzl-oh,s-2-amino-3-benzyloxy propanoic acid,l-serine, o-phenylmethyl,2s-2-amino-3-benzyloxy propanoic acid,o-phenylmethyl-l-serine,serine, o-phenylmethyl,benzylserine,o-benzylserine #,z-o-benzyl-l-serine
78457
(2S)-2-amino-3-phenylmethoxypropanoic acid
C1=CC=C(C=C1)COCC(C(=O)O)N
4726-96-9
C10H13NO3
195.22
IDGQXGPQOGUGIX-VIFPVBQESA-N
o-benzyl-l-serine,h-ser bzl-oh,s-2-amino-3-benzyloxy propanoic acid,l-serine, o-phenylmethyl,2s-2-amino-3-benzyloxy propanoic acid,o-phenylmethyl-l-serine,serine, o-phenylmethyl,benzylserine,o-benzylserine #,z-o-benzyl-l-serine
78457
(2S)-2-amino-3-phenylmethoxypropanoic acid
C1=CC=C(C=C1)COCC(C(=O)O)N
Specific Rotation 22.5
Melting Point 210°C to 213°C
Packaging Glass bottle
Water 1% Max.
Quantity 25g
Color White
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (HPLC)
Formula Weight 195.22
Physical Form Powder
Percent Purity 99%
Chemical Name or Material O-Benzyl-L-serine
Show More Show Less

EINECSNumber : 225-220-6