Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ O-Benzyl-L-threonine hydrochloride, 95%

Click to view available options
Quantity:
1g
5g
60856-51-1
C11H16ClNO3
245.703
MFCD00037768
XECHCUXLJZQHOJ-SCYNACPDSA-N
h-thr bzl-oh hcl,h-thr bzl-oh.hcl,2s,3r-2-amino-3-benzyloxy butanoic acid hydrochloride,o-benzyl-l-threonine,h-thr bzl-oh??hcl,h-thr bzl-ohhcl,h-thr bzl-oh hydrochloride,l-threonine,o-phenylmethyl-,hydrochloride 9ci
13919385
(2S,3R)-2-amino-3-phenylmethoxybutanoic acid;hydrochloride
CC(C(C(=O)O)N)OCC1=CC=CC=C1.Cl
60856-51-1
C11H16ClNO3
245.703
MFCD00037768
XECHCUXLJZQHOJ-SCYNACPDSA-N
h-thr bzl-oh hcl,h-thr bzl-oh.hcl,2s,3r-2-amino-3-benzyloxy butanoic acid hydrochloride,o-benzyl-l-threonine,h-thr bzl-oh??hcl,h-thr bzl-ohhcl,h-thr bzl-oh hydrochloride,l-threonine,o-phenylmethyl-,hydrochloride 9ci
13919385
(2S,3R)-2-amino-3-phenylmethoxybutanoic acid;hydrochloride
CC(C(C(=O)O)N)OCC1=CC=CC=C1.Cl
Formula Weight 245.7
Physical Form Solid
Percent Purity 95%
Quantity 5g
Chemical Name or Material O-Benzyl-L-threonine hydrochloride

storageNote1 : 2° to 8°C