Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ O-Benzyl-N-benzyloxycarbonyl-L-threonine, 95%

Click to view available options
Quantity:
1g
5g
25g
69863-36-1
C19H21NO5
343.379
MFCD00077042
FVKJXAYBJIAXAU-PBHICJAKSA-N
z-thr bzl-oh,n-z-o-benzyl-l-threonine,n-cbz-o-benzyl-l-threonine,cbz-thr bzl-oh,o-benzyl-n-benzyloxycarbonyl-l-threonine,2s,3r-3-benzyloxy-2-benzyloxy carbonyl amino butanoic acid,2s,3r-3-benzyloxy-2-benzyloxy carbonyl amino butanoic acid,ambotzzaa1248,pubchem19048,z-o-benzyl-l-threonine
7018810
(2S,3R)-3-phenylmethoxy-2-(phenylmethoxycarbonylamino)butanoic acid
CC(C(C(=O)O)NC(=O)OCC1=CC=CC=C1)OCC2=CC=CC=C2
69863-36-1
C19H21NO5
343.379
MFCD00077042
FVKJXAYBJIAXAU-PBHICJAKSA-N
z-thr bzl-oh,n-z-o-benzyl-l-threonine,n-cbz-o-benzyl-l-threonine,cbz-thr bzl-oh,o-benzyl-n-benzyloxycarbonyl-l-threonine,2s,3r-3-benzyloxy-2-benzyloxy carbonyl amino butanoic acid,2s,3r-3-benzyloxy-2-benzyloxy carbonyl amino butanoic acid,ambotzzaa1248,pubchem19048,z-o-benzyl-l-threonine
7018810
(2S,3R)-3-phenylmethoxy-2-(phenylmethoxycarbonylamino)butanoic acid
CC(C(C(=O)O)NC(=O)OCC1=CC=CC=C1)OCC2=CC=CC=C2
Beilstein 5988744
Formula Weight 343.38
Physical Form Solid
Percent Purity 95%
Melting Point 77°C to 80°C
Quantity 1g
Chemical Name or Material O-Benzyl-N-benzyloxycarbonyl-L-threonine