Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-Benzylhydroxylamine, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
622-33-3
C7H9NO
123.15
MFCD00221709
XYEOALKITRFCJJ-UHFFFAOYSA-N
benzyloxyamine,hydroxylamine, o-phenylmethyl,hydroxylamine, o-benzyl,o-benzyl-hydroxylamine,benzyl-o-hydroxylamine,oxybenzylamine,o-phenylmethyl hydroxylamine,o-benzyloxylamine,phenylmethoxyamine,benzyloxy amine
102313
O-benzylhydroxylamine
C1=CC=C(C=C1)CON
622-33-3
C7H9NO
123.15
MFCD00221709
XYEOALKITRFCJJ-UHFFFAOYSA-N
benzyloxyamine,hydroxylamine, o-phenylmethyl,hydroxylamine, o-benzyl,o-benzyl-hydroxylamine,benzyl-o-hydroxylamine,oxybenzylamine,o-phenylmethyl hydroxylamine,o-benzyloxylamine,phenylmethoxyamine,benzyloxy amine
102313
O-benzylhydroxylamine
C1=CC=C(C=C1)CON
Boiling Point 110°C to 112°C (20.0mmHg)
Specific Gravity 1.19
Infrared Spectrum Authentic
Refractive Index 1.5390 to 1.5410 (20°C, 589nm)
Quantity 100g
Density 1.1900g/mL
Packaging Glass bottle
Color Colorless
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula C6H5CH2ONH2
Beilstein 06, IV, 2562
Solubility Information Solubility in water: soluble
Formula Weight 123.15
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material O-Benzylhydroxylamine
Show More Show Less

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning