Learn More
Sklar InstrumentsSupplier Diversity Partner O'Brien Stitch Scissors, Sklar, 3-3/4 in., Sharp/Sharp, Scissors, Angled, Smooth, Finger Rings, Stainless Steel.EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 222837
O'Brien Stitch Scissors, Sklar, 3-3/4 in., Sharp/Sharp, Scissors, Angled, Smooth, Finger Rings, Stainless Steel. Sklar O'Brien Stitch Scissors are primarily used for suture removal. These scissors have sharp, fine, angled tips that are used to snip the suture rather than lifting the suture slightly first, as with Spencer or Littauer stitch scissors. They are often used in dental procedures where lifting the suture may be unpractical due to location and space limitations. This product is angled with smooth, sharp/sharp tips and a length of 3-3/4 inches.
Catalog No. 50-143-7859