Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromophenoxyacetic acid, 98%, Acros Organics

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
1879-56-7
C8H7BrO3
231.05
MFCD00234407
AZYODYPUWJPKOI-UHFFFAOYSA-N
2-bromophenoxy acetic acid,2-2-bromophenoxy acetic acid,o-bromophenoxyacetic acid,2-bromo-phenoxy-acetic acid,2-bromophenoxyacetic acid,acetic acid, 2-bromophenoxy,acetic acid,2-2-bromophenoxy,2-bromo phenoxyacetic acid,2-bromophenoxy-acetic acid
592201
2-(2-bromophenoxy)acetic acid
C1=CC=C(C(=C1)OCC(=O)O)Br
1879-56-7
C8H7BrO3
231.05
MFCD00234407
AZYODYPUWJPKOI-UHFFFAOYSA-N
2-bromophenoxy acetic acid,2-2-bromophenoxy acetic acid,o-bromophenoxyacetic acid,2-bromo-phenoxy-acetic acid,2-bromophenoxyacetic acid,acetic acid, 2-bromophenoxy,acetic acid,2-2-bromophenoxy,2-bromo phenoxyacetic acid,2-bromophenoxy-acetic acid
592201
2-(2-bromophenoxy)acetic acid
C1=CC=C(C(=C1)OCC(=O)O)Br
Packaging Glass bottle
Melting Point 141°C to 145°C
Color Pink to White
Quantity 25g
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula BrC6H4OCH2CO2H
Formula Weight 231.05
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 2-Bromophenoxyacetic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning