Alfa Aesar™ o-Carborane, 98%

Manufacturer: Alfa Aesar™ 35131ME

Quantity 500mg

 View more versions of this product

Catalog No. AA35131ME

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 16872-09-6
    Formula Weight 144.25
    MDL Number MFCD00012169
    UN Number UN1325
    Synonym 1,2-Dicarbadodecaborane
    Chemical Name or Material o-Carborane

    Specifications

    Formula Weight 144.25
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P241-P301+P312-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    Use explosion-proof electrical/ventilating/lighting/ equipment.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Density 0.95g/mL
    MDL Number MFCD00012169
    UN Number UN1325
    Synonym 1,2-Dicarbadodecaborane
    CAS 16872-09-6
    Color White
    TSCA TSCA
    Physical Form Powder
    Solubility Information Insoluble in water
    Health Hazard 1 Warning
    Melting Point 285° to 287°C
    Quantity 500mg
    Health Hazard 2 GHS H Statement
    H228-H302
    Flammable solid.
    Harmful if swallowed.
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
    RTECS Number HS9285000
    Chemical Name or Material o-Carborane
    Assay 98%

    GHS H Statement
    H228-H302
    Flammable solid.
    Harmful if swallowed.

    GHS P Statement
    P210-P280-P240-P241-P301+P312-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    Use explosion-proof electrical/ventilating/lighting/ equipment.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit