Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ o-Cresolphthalein complexone tetrasodium salt

Click to view available options
Quantity:
1g
5g
25g
62698-54-8
C32H28N2Na4O12
724.537
MFCD00066371
KSMPPVYJZJBIES-UHFFFAOYSA-J
phthalein complexon sodium salt,o-cresolphthalein complexone tetrasodium salt,sodium 2,2',2,2'-3-oxo-1,3-dihydroisobenzofuran-1,1-diyl bis 2-hydroxy-3-methyl-5,1-phenylene bis methylene bis azanetriyl tetraacetate,phthaleincomplexonsodiumsalt,phthalein complexone sodium salt,glycine, n,n'-3-oxo-1 3h-isobenzofuranylidene bis 6-hydroxy-5-methyl-3,1-phenylene methylene bis n-carboxymethyl-, sodium salt 1:4,glycine, n,n'-3-oxo-1 3h-isobenzofuranylidene bis 6-hydroxy-5-methyl-3,1-phenylene methylene bis n-carboxymethyl-, tetrasodium salt,tetrasodium 2,2',2,2'-3-oxo-1,3-dihydro-2-benzofuran-1,1-diyl bis 6-hydroxy-5-methylbenzene-3,1-diyl methanediylnitrilo tetraacetate,tetrasodium 2-5-1-3-bis carboxylatomethyl amino methyl-4-hydroxy-5-methylphenyl-3-oxo-2-benzofuran-1-yl-2-hydroxy-3-methylphenyl methyl carboxylatomethyl amino acetate,tetrasodium 2-5-1-3-bis 2-oxido-2-oxoethyl amino methyl-4-hydroxy-5-methylphenyl-3-oxo-2-benzofuran-1-yl-2-hydroxy-3-methylphenyl methyl-2-oxido-2-oxoethyl amino acetate
112916
tetrasodium;2-[[5-[1-[3-[[bis(carboxylatomethyl)amino]methyl]-4-hydroxy-5-methylphenyl]-3-oxo-2-benzofuran-1-yl]-2-hydroxy-3-methylphenyl]methyl-(carboxylatomethyl)amino]acetate
CC1=C(C(=CC(=C1)C2(C3=CC=CC=C3C(=O)O2)C4=CC(=C(C(=C4)CN(CC(=O)[O-])CC(=O)[O-])O)C)CN(CC(=O)[O-])CC(=O)[O-])O.[Na+].[Na+].[Na+].[Na+]
62698-54-8
C32H28N2Na4O12
724.537
MFCD00066371
KSMPPVYJZJBIES-UHFFFAOYSA-J
phthalein complexon sodium salt,o-cresolphthalein complexone tetrasodium salt,sodium 2,2',2,2'-3-oxo-1,3-dihydroisobenzofuran-1,1-diyl bis 2-hydroxy-3-methyl-5,1-phenylene bis methylene bis azanetriyl tetraacetate,phthaleincomplexonsodiumsalt,phthalein complexone sodium salt,glycine, n,n'-3-oxo-1 3h-isobenzofuranylidene bis 6-hydroxy-5-methyl-3,1-phenylene methylene bis n-carboxymethyl-, sodium salt 1:4,glycine, n,n'-3-oxo-1 3h-isobenzofuranylidene bis 6-hydroxy-5-methyl-3,1-phenylene methylene bis n-carboxymethyl-, tetrasodium salt,tetrasodium 2,2',2,2'-3-oxo-1,3-dihydro-2-benzofuran-1,1-diyl bis 6-hydroxy-5-methylbenzene-3,1-diyl methanediylnitrilo tetraacetate,tetrasodium 2-5-1-3-bis carboxylatomethyl amino methyl-4-hydroxy-5-methylphenyl-3-oxo-2-benzofuran-1-yl-2-hydroxy-3-methylphenyl methyl carboxylatomethyl amino acetate,tetrasodium 2-5-1-3-bis 2-oxido-2-oxoethyl amino methyl-4-hydroxy-5-methylphenyl-3-oxo-2-benzofuran-1-yl-2-hydroxy-3-methylphenyl methyl-2-oxido-2-oxoethyl amino acetate
112916
tetrasodium;2-[[5-[1-[3-[[bis(carboxylatomethyl)amino]methyl]-4-hydroxy-5-methylphenyl]-3-oxo-2-benzofuran-1-yl]-2-hydroxy-3-methylphenyl]methyl-(carboxylatomethyl)amino]acetate
CC1=C(C(=CC(=C1)C2(C3=CC=CC=C3C(=O)O2)C4=CC(=C(C(=C4)CN(CC(=O)[O-])CC(=O)[O-])O)C)CN(CC(=O)[O-])CC(=O)[O-])O.[Na+].[Na+].[Na+].[Na+]
Quantity 5g
Formula Weight 724.54
Chemical Name or Material o-Cresolphthalein complexone tetrasodium salt

EINECSNumber : 305-335-9

TSCA : Yes

Recommended Storage : Ambient temperatures