o-Dianisidine, >98%, Alfa Aesar™

Manufacturer: Alfa Aesar A1715006

Quantity 5g

 View more versions of this product

Catalog No. AAA1715006

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >98%
    Molecular Formula C14H16N2O2
    Formula Weight 244.3
    MDL Number MFCD00008372
    Synonym 3, 3′-Dimethoxybenzidine
    Chemical Name or Material o-Dianisidine

    Specifications

    Formula Weight 244.3
    Quantity 5g
    TSCA TSCA
    Beilstein 1879884
    Merck Index 14,2991
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H350-H302
    May cause cancer.
    Harmful if swallowed.
    Health Hazard 3 GHS P Statement
    P281-P264-P301+P312-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Wash thoroughly after handling.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    IF exposed or con
    RTECS Number DD0875000
    EINECS Number 204-355-4
    Assay Percent Range >98%
    Molecular Formula C14H16N2O2
    MDL Number MFCD00008372
    CAS 119-90-4
    Synonym 3, 3′-Dimethoxybenzidine
    Chemical Name or Material o-Dianisidine
    Assay ≥98%

    Description

    Applications
    o-Dianisidine is used as a redox indicator. It is used as an intermediate for the preparation of azo dyes, pigments and o-dianisidine diisocyanate, which finds application in adhesives, polyurethane elastomers and resins. It is also used in various industries like leather, paper, plastics, rubber, and textiles. It acts as a reagent to detect metals, thiocyanates, and nitrites.

    Solubility
    Soluble in alcohol, benzene, ether, chloroform, acetone, most organic solvents and lipids. Slightly soluble in water.

    Notes
    Incompatible with strong oxidizing agents.

    GHS H Statement
    H350-H302
    May cause cancer.
    Harmful if swallowed.

    GHS P Statement
    P281-P264-P301+P312-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Wash thoroughly after handling.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    IF exposed or con

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit