Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Dichlorobenzene (Technical), Fisher Chemical

Catalog Number b2484
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
95-50-1
C6H4Cl2
146.998
MFCD00000535
RFFLAFLAYFXFSW-UHFFFAOYSA-N
o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben
7239
CHEBI:35290
1,2-dichlorobenzene
C1=CC=C(C(=C1)Cl)Cl
This item is not returnable. View return policy
95-50-1
C6H4Cl2
146.998
MFCD00000535
RFFLAFLAYFXFSW-UHFFFAOYSA-N
o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben
7239
CHEBI:35290
1,2-dichlorobenzene
C1=CC=C(C(=C1)Cl)Cl
Alpha Vector Dichlorobenzene
Melting Point -15°C
Color Yellow
Density 1.30g/cm³
Boiling Point 179°C
Packaging Glass Bottle
Refractive Index 1.5481 to 1.5501 (at 25°C)
Quantity 4 L
UN Number 1591
Vapor Density 5.05g/mol
Vapor Pressure 1.15mmHg at 20°C
Formula Weight 147
Grade Technical
Physical Form Liquid
Chemical Name or Material o-Dichlorobenzene
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit