Alfa Aesar™ O-Ethoxy-L-serine hydrochloride, 97%

Manufacturer: Alfa Aesar™ H5279003

Quantity 1g

 View more versions of this product

Catalog No. AAH5279003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4775-82-0
    Molecular Formula C5H11NO3·HCl
    Formula Weight 169.61
    MDL Number MFCD06797557
    Synonym (S)-2-Amino-3-ethoxypropionic acid hydrochloride
    Chemical Name or Material O-Ethoxy-L-serine hydrochloride

    Specifications

    Formula Weight 169.61
    Molecular Formula C5H11NO3·HCl
    CAS 4775-82-0
    MDL Number MFCD06797557
    Synonym (S)-2-Amino-3-ethoxypropionic acid hydrochloride
    Quantity 1g
    Chemical Name or Material O-Ethoxy-L-serine hydrochloride
    Assay 97%

    Description

    Applications
    O-Ethoxy-L-serine hydrochloride is used as a pharmaceutical intermediate.

    Solubility
    Soluble in water, ethanol.

    Notes
    Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit