Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Fluorotoluene, 99%, Acros Organics™

Click to view available options
Quantity:
10g
100g
500g
2.5kg
Packaging:
Glass bottle
95-52-3
C7H7F
110.13
MFCD00000322
MMZYCBHLNZVROM-UHFFFAOYSA-N
2-fluorotoluene,o-fluorotoluene,benzene, 1-fluoro-2-methyl,toluene, o-fluoro,1-methyl-2-fluorobenzene,unii-l55xkb3k7h,1-fluoro-2-methyl-benzene,ortho-fluorotoluene,fluorotoluene,l55xkb3k7h
7241
1-fluoro-2-methylbenzene
CC1=CC=CC=C1F
95-52-3
C7H7F
110.13
MFCD00000322
MMZYCBHLNZVROM-UHFFFAOYSA-N
2-fluorotoluene,o-fluorotoluene,benzene, 1-fluoro-2-methyl,toluene, o-fluoro,1-methyl-2-fluorobenzene,unii-l55xkb3k7h,1-fluoro-2-methyl-benzene,ortho-fluorotoluene,fluorotoluene,l55xkb3k7h
7241
1-fluoro-2-methylbenzene
CC1=CC=CC=C1F
Specific Gravity 1
Color Colorless to Yellow
Melting Point -62.0°C
Infrared Spectrum Authentic
Refractive Index 1.4720 to 1.4750
Quantity 100g
Packaging Glass bottle
Flash Point 8°C
Boiling Point 113.0°C to 114.0°C
Density 1.0000g/mL
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula CH3C6H4F
Beilstein 05, 290
Merck Index 15, 4212
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol and ether
Formula Weight 110.13
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material o-Fluorotoluene, 99+%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 202-428-5

RTECSNumber : XT2579000

TSCA : TSCA