Learn More
Thermo Scientific™ O'GeneRuler™ Ready-to-use 100bp Plus DNA Ladder: PROMO
Perform sizing and approximate quantification of a wide range of double-stranded DNA on agarose or polyacrylamide gels.
Manufacturer:  Thermo Scientific™ SM1143
 View more versions of this product
Catalog No. FERSM1143PM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
O'GeneRuler 100 bp Plus DNA Ladder, ready-to-use, is recommended for sizing and approximate quantification of a wide range of double-stranded DNA on agarose or polyacrylamide gels. The ladder is a mixture of chromatography-purified individual DNA fragments. O'GeneRuler 100 bp Plus DNA Ladder, ready-to-use, is premixed with 6X Orange DNA Loading Dye for direct loading on gel. 6X Orange DNA Loading Dye contains xylene cyanol FF and orange G. In a 1% agarose gel orange G dye migrates at 50 bp. The ladder is supplied with 6X Orange DNA Loading Dye for sample DNA.
  • Ideal for both DNA sizing and approximate quantification
  • Sharp bands
  • Bright reference bands (given in red)
  • Can be directly loaded and are stable at room temperature for 6 months
  • Supplied with loading dye for sample DNA
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
This product or its use is covered by at least one claim of U.S. Patent Nos. 5,834,201, 6,680,378, and/or 7,132,520 owned by Invitrogen Corporation. The purchase of this product conveys to the buyer the non-transferable right to use the purchased amount