Learn More
O-GlcNAc Mouse anti-All, Human, PE, Clone: RL2, eBioscience™, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
 View more versions of this product
Catalog No. 12-979-341
Add to cart
Description
Description
Mouse Monoclonal Antibody
O-GlcNAc Monoclonal antibody specifically detects O-GlcNAc in All, Human samples. It is validated for Flow Cytometry
Specifications
Specifications
O-GlcNAc
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
O15294
1110038P24Rik; 4831420N21Rik; AI115525; GlcNAc; HINCUT-1; HRNT1; O-GLCNAC; O-GlcNAc transferase subunit p110; Ogt; Ogtl; O-linked N-acetylglucosamine transferase 110 kDa subunit; UDP-N-acetylglucosamine--peptide N-acetylglucosaminyltransferase 110 kDa subunit
Mouse
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
8473
Flow Cytometry
RL2
PE
OGT
Liquid
OGT
Rat Liver nuclear envelopes
Affinity chromatography
RUO
Antibody
Monoclonal
All, Human