Learn More
O-GlcNAc Mouse anti-Non-species specific, DyLight 405, Clone: 9B9, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259309V
Catalog No. NBP259309V
Add to cart
Description
Description
O-GlcNAc Monoclonal antibody specifically detects O-GlcNAc in Non-species specific samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
O-GlcNAc
9B9
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Purified
Mouse
IgM
100 ul
Primary
Specific for B-O-Linked N-Acetylgalactosamine and B-O-Linked N-Acetylglucosamine modified proteins. Does not cross-react with Glucose or Galactose modified proteins.
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
GlcNAc, O-linked N-acetylglucosamine
Synthetic B-O-Linked N-Acetylglucosamine conjugated to Keyhole Limpet Hemocyanin (KLH).
PEG purified
Store at 4C in the dark.
Monoclonal
Non-species specific