Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ O-Methyl-D-serine hydrochloride, 95%

Click to view available options
Quantity:
250mg
1g
5g
86118-10-7
C4H10ClNO3
155.578
MFCD11099371
NLDVBPFPOCWZCE-AENDTGMFSA-N
r-2-amino-3-methoxypropanoic acid hydrochloride,o-methyl-d-serine hydrochloride,2r-2-amino-3-methoxypropanoic acid hydrochloride,d-serine, o-methyl-, hydrochloride,o-methyl-d-ser hcl,d-serine, o-methyl-, hydrochloride 1:1,ksc497q9r,o-methyl-d-serine-hydrogen chloride 1/1,r-2-amino-3-methoxypropanoic acid hcl,r-2-amino-3-methoxypropanoicacidhydrochloride
56965747
(2R)-2-amino-3-methoxypropanoic acid;hydrochloride
COCC(C(=O)O)N.Cl
86118-10-7
C4H10ClNO3
155.578
MFCD11099371
NLDVBPFPOCWZCE-AENDTGMFSA-N
r-2-amino-3-methoxypropanoic acid hydrochloride,o-methyl-d-serine hydrochloride,2r-2-amino-3-methoxypropanoic acid hydrochloride,d-serine, o-methyl-, hydrochloride,o-methyl-d-ser hcl,d-serine, o-methyl-, hydrochloride 1:1,ksc497q9r,o-methyl-d-serine-hydrogen chloride 1/1,r-2-amino-3-methoxypropanoic acid hcl,r-2-amino-3-methoxypropanoicacidhydrochloride
56965747
(2R)-2-amino-3-methoxypropanoic acid;hydrochloride
COCC(C(=O)O)N.Cl
Beilstein 56965747
Formula Weight 155.58
Physical Form Solid
Percent Purity 95%
Quantity 250mg
Sensitivity Hygroscopic
Chemical Name or Material O-Methyl-D-serine hydrochloride