Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ O-Methylisourea hemisulfate, 99%

Click to view available options
Quantity:
50g
250g
1000g
52328-05-9
C4H14N4O6S
246.238
MFCD00040594
QSCPQKVWSNUJLJ-UHFFFAOYSA-N
o-methylisourea hemisulfate,2-methylisouronium sulfate,o-methylisourea hemisulfate salt,bis methoxymethanimidamide ; sulfuric acid,carbamimidic acid, methyl ester, sulfate 2:1,omi™,o-methylpseudourea sulfate,0-methylisourea hemisulfate,o-methylisourea hemisulphate,bis 2-methylisouronium sulphate
11357018
methyl carbamimidate;sulfuric acid
COC(=N)N.COC(=N)N.OS(=O)(=O)O
52328-05-9
C4H14N4O6S
246.238
MFCD00040594
QSCPQKVWSNUJLJ-UHFFFAOYSA-N
o-methylisourea hemisulfate,2-methylisouronium sulfate,o-methylisourea hemisulfate salt,bis methoxymethanimidamide ; sulfuric acid,carbamimidic acid, methyl ester, sulfate 2:1,omi™,o-methylpseudourea sulfate,0-methylisourea hemisulfate,o-methylisourea hemisulphate,bis 2-methylisouronium sulphate
11357018
methyl carbamimidate;sulfuric acid
COC(=N)N.COC(=N)N.OS(=O)(=O)O
Applications
O-Methylisoureahemisulfate is used for the synthesis of fluorouracil antineoplastic and anthelmintic imidazole.

Solubility
Soluble in organic solvents, water, methanol and ethanol

Notes
Incompatible with strong oxidizing agents.
Quantity 250g
Melting Point 163°C to 167°C
Formula Weight 123.12
Percent Purity 99%
Chemical Name or Material O-Methylisourea hemisulfate