Alfa Aesar™ O-Methylisourea hydrochloride, 98%

Manufacturer: Alfa Aesar™ A1477309

Quantity 10g

 View more versions of this product

Catalog No. AAA1477309

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5329-33-9
    Molecular Formula C2H6N2O·HCl
    Formula Weight 110.54
    MDL Number MFCD00035043
    Beilstein 3909619
    Chemical Name or Material O-Methylisourea hydrochloride

    Specifications

    Formula Weight 110.54
    Molecular Formula C2H6N2O·HCl
    MDL Number MFCD00035043
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Melting Point ≈116°C (dec.)
    Quantity 10g
    TSCA TSCA
    CAS 5329-33-9
    Beilstein 3909619
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material O-Methylisourea hydrochloride
    Assay 98%

    Description

    Applications
    O-methylisourea hydrochloride is used to prepare 1-chloro-2-methyl-isourea. It is also used as an organic intermediate.

    Solubility
    Soluble in organic solvents, water, methanol and ethanol.

    Notes
    Incompatible with strong oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit