Alfa Aesar™ O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium hexafluorophosphate, 98+%

Manufacturer: Alfa Aesar™ H5403503

Quantity 1g

 View more versions of this product

Catalog No. AAH5403503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 265651-18-1
    Molecular Formula C9H16F6N3O3P
    Formula Weight 359.21
    MDL Number MFCD01863753
    Synonym N,N,N',N'-Tetramethyl-O-(N-succinimidyl)uronium hexafluorophosphate; HSTU
    Chemical Name or Material O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium hexafluorophosphate

    Specifications

    Formula Weight 359.21
    Molecular Formula C9H16F6N3O3P
    MDL Number MFCD01863753
    CAS 265651-18-1
    Synonym N,N,N',N'-Tetramethyl-O-(N-succinimidyl)uronium hexafluorophosphate; HSTU
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point ≈220°C
    Quantity 1g
    Recommended Storage −30° to −10°C
    Chemical Name or Material O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium hexafluorophosphate
    Assay ≥98%

    Description

    Applications
    Coupling reagent for peptide synthesis and the formation of other amides. Reactant for synthesis of liposomal contrast agents for magnetic resonance imaging, synthesis of protein labeling molecules, synthesis of thiol-reactive Cy5 derivatives.

    Solubility
    Soluble in acetonitrile 0.5 g/mL.

    Notes
    Moisture sensitive. Store under inert gas. Store away from oxidizing agents, water/ moisture.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    −30° to −10°C

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit