Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Nitroacetophenone 95%, ACROS Organics™

Click to view available options
Quantity:
2.5g
20g
Packaging:
Glass bottle
577-59-3
C8H7NO3
165.15
MFCD00007144
SUGXZLKUDLDTKX-UHFFFAOYSA-N
2'-nitroacetophenone,1-2-nitrophenyl ethanone,o-nitroacetophenone,ethanone, 1-2-nitrophenyl,2-acetylnitrobenzene,acetophenone, 2'-nitro,methyl 2-nitrophenyl ketone,unii-52l53k4x3o,ccris 2329,1-2-nitrophenyl ethan-1-one
11346
1-(2-nitrophenyl)ethanone
CC(=O)C1=CC=CC=C1[N+](=O)[O-]
577-59-3
C8H7NO3
165.15
MFCD00007144
SUGXZLKUDLDTKX-UHFFFAOYSA-N
2'-nitroacetophenone,1-2-nitrophenyl ethanone,o-nitroacetophenone,ethanone, 1-2-nitrophenyl,2-acetylnitrobenzene,acetophenone, 2'-nitro,methyl 2-nitrophenyl ketone,unii-52l53k4x3o,ccris 2329,1-2-nitrophenyl ethan-1-one
11346
1-(2-nitrophenyl)ethanone
CC(=O)C1=CC=CC=C1[N+](=O)[O-]
Quantity 2.5g
Packaging Glass bottle
Boiling Point 159.0°C (16.0 mmHg)
Melting Point 28.0°C to 30.0°C
Flash Point >112°C
Refractive Index 1.5500 to 1.5520
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula O2NC6H4COCH3
Beilstein 07, 288
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol and ether
Formula Weight 165.15
Percent Purity 95%
Chemical Name or Material o-Nitroacetophenone, 95%
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 209-414-8

RTECSNumber : AM9600000

TSCA : TSCA