Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Nitrocinnamaldehyde 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
1466-88-2
C9H7NO3
177.16
MFCD00007188
VMSMELHEXDVEDE-HWKANZROSA-N
2-nitrocinnamaldehyde,3-2-nitrophenyl acrylaldehyde,o-nitrocinnamaldehyde,trans-2-nitrocinnamaldehyde,2e-3-2-nitrophenyl prop-2-enal,2-propenal, 3-2-nitrophenyl,e-3-2-nitrophenyl acrylaldehyde,e-3-2-nitrophenyl prop-2-enal,3-2-nitrophenyl prop-2-enal,ccris 3775
5367122
(E)-3-(2-nitrophenyl)prop-2-enal
C1=CC=C(C(=C1)C=CC=O)[N+](=O)[O-]
1466-88-2
C9H7NO3
177.16
MFCD00007188
VMSMELHEXDVEDE-HWKANZROSA-N
2-nitrocinnamaldehyde,3-2-nitrophenyl acrylaldehyde,o-nitrocinnamaldehyde,trans-2-nitrocinnamaldehyde,2e-3-2-nitrophenyl prop-2-enal,2-propenal, 3-2-nitrophenyl,e-3-2-nitrophenyl acrylaldehyde,e-3-2-nitrophenyl prop-2-enal,3-2-nitrophenyl prop-2-enal,ccris 3775
5367122
(E)-3-(2-nitrophenyl)prop-2-enal
C1=CC=C(C(=C1)C=CC=O)[N+](=O)[O-]
Infrared Spectrum Authentic
Color Brown-Yellow to Yellow
Melting Point 125°C to 129°C
Quantity 5g
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula O2NC6H4CH=CHCHO
Beilstein 07, 358
Solubility Information Solubility in water: slightly soluble
Formula Weight 177.16
Physical Form Crystals or Powder
Percent Purity ≥97.5%
Chemical Name or Material o-Nitrocinnamaldehyde, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

AIR SENSITIVE

EINECSNumber : 215-988-0