Alfa Aesar™ (-)-O,O'-Dipivaloyl-L-tartaric acid, 98+%

Manufacturer: Alfa Aesar™ H5603206

Quantity 5g

 View more versions of this product

Catalog No. AAH5603206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 65259-81-6
    Molecular Formula C14H22O8
    Formula Weight 318.32
    MDL Number MFCD00066535
    Beilstein 2008171
    Synonym (-)-DPTA
    Chemical Name or Material (-)-O,O'-Dipivaloyl-L-tartaric acid

    Specifications

    Formula Weight 318.32
    Melting Point 127° to 132°C
    Quantity 5g
    Optical Rotation −24° (c=1.7 in Dioxane)
    Molecular Formula C14H22O8
    MDL Number MFCD00066535
    CAS 65259-81-6
    Beilstein 2008171
    Synonym (-)-DPTA
    Chemical Name or Material (-)-O,O'-Dipivaloyl-L-tartaric acid
    Assay ≥98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit