Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Orsellinic acid monohydrate, 98%, Acros Organics™

Click to view available options
Quantity:
1g
5g
480-64-8
C8H10O5
186.17
AMKYESDOVDKZKV-UHFFFAOYSA-N
orsellinic acid,orsellic acid,o-orsellinic acid,orcinolcarboxylic acid,4,6-dihydroxy-o-toluic acid,unii-11xla0494b,benzoic acid, 2,4-dihydroxy-6-methyl,2,4-dihydroxy-6-methyl-benzoic acid,orsellinate,4,6-dihydroxy-2-methylbenzoic acid
68072
CHEBI:32807
2,4-dihydroxy-6-methylbenzoic acid
CC1=CC(=CC(=C1C(=O)O)O)O
480-64-8
C8H10O5
186.17
AMKYESDOVDKZKV-UHFFFAOYSA-N
orsellinic acid,orsellic acid,o-orsellinic acid,orcinolcarboxylic acid,4,6-dihydroxy-o-toluic acid,unii-11xla0494b,benzoic acid, 2,4-dihydroxy-6-methyl,2,4-dihydroxy-6-methyl-benzoic acid,orsellinate,4,6-dihydroxy-2-methylbenzoic acid
68072
CHEBI:32807
2,4-dihydroxy-6-methylbenzoic acid
CC1=CC(=CC(=C1C(=O)O)O)O
Packaging Glass Bottle
Water 10 % max.
Quantity 1g
Infrared Spectrum Authentic
Melting Point 152.0°C to 153.0°C
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 186.17
Percent Purity 98%
Chemical Name or Material o-Orsellinic acid monohydrate

GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Call a POISON CENTER or doctor/physician if you feel unwell. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning