Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
o-Phosphoric Acid, 85% (Certified ACS), Fisher Chemical™
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
Packaging:
Flint Glass Bottle
Glass Bottle
Plastic Bottle
Safe-Cote™ Glass
Specifications
pH | 1 |
Melting Point | 21°C |
Boiling Point | 158°C |
Density | 1.680g/cm³ |
Heavy Metals (as Pb) | 0.001% max. |
Identification | Pass Test |
Packaging | Safe-Cote™ Glass |
Reducing Substances | Pass Test |
Vapor Pressure | 2 hPa at 20°C |
Normality | about 44.6 |
Show More |
Safety and Handling
Hazard Statement | DANGER! |
Precautionary Statement | Emergency Overview Causes burns by all exposure routes. Use only under a chemical fume hood. Wear personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Do not induce vomiting. Do not induce vomiting. Call a physician or Poison Control Center immediately. NFPA Health:3 Flammability:0 Instability:0 |
DOTInformation | DOT Class 8, : Corrosive |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction