o-Phosphoric Acid, 85% (Certified ACS), Fisher Chemical

Packaging Poly Bottle
Quantity 500mL

 View more versions of this product

Catalog No. A242P-500

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  This product cannot be ordered online, as it can only be purchased internationally. 

Custom packing/Bulk ordering

You can now order this item in custom quantities.  Learn More

    Chemical Identifiers

    CAS 7664-38-2,7732-18-5
    Assay Percent Range ≥85.0 w/w %
    Molecular Formula H3O4P
    Formula Weight 98
    MDL Number MFCD00011340
    Synonym Orthophosphoric Acid
    Chemical Name or Material o-Phosphoric Acid, 85%
    Assay ≥85.0% (w/w)
    Grade Certified ACS Grade

    Specifications

    Melting Point 21°C
    Boiling Point 158°C
    Formula Weight 98
    Quantity 500mL
    DOT Information DOT Class 8, : Corrosive
    Physical Form Liquid
    pH 1
    Color Clear Viscous
    Alpha Vector Phosphoric Acid
    Normality about 44.6
    Density 1.69g/mL
    Vapor Pressure 2 hPa at 20°C
    Health Hazard 3 Emergency Overview
    Causes burns by all exposure routes. Use only under a chemical fume hood. Wear personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Do not induce vomiting. Do not induce vomiting. Call a physician or Poison Control Center immediately.

    NFPA

    Health:3
    Flammability:0
    Instability:0
    Potassium (K) 0.005% max.
    Sodium (Na) 0.025% max.
    Sulfate 0.001% max.
    Heavy Metals (as Pb) 0.001% max.
    Identification Pass Test
    Phosphorous and Hypophosphorous Acid Pass Test
    Arsenic (As) 0.5ppm max.
    Calcium (Ca) 0.002% max.
    MDL Number MFCD00011340
    Chloride 3ppm max.
    Additional Information Vapor Pressure: 2hPa at 20°C
    Iron (Fe) 0.003% max.
    Magnesium (Mg) 0.002% max.
    Reducing Substances Pass Test
    Reaction with H2SO4 Pass Test
    Volatile Acids 0.001% max.
    Assay Percent Range ≥85.0 w/w %
    Synonym Orthophosphoric Acid
    Antimony (Sb) 0.002% max.
    CAS 7732-18-5,7664-38-2
    Insoluble Matter 0.001% max.
    Manganese (Mn) 0.5ppm max.
    Nitrate 5ppm max.
    Packaging Poly Bottle
    Cleanup Symbol Acid
    Molecular Formula H3O4P
    ChemAlert Storage Symbol White
    Health Hazard 2 DANGER!
    Chemical Name or Material o-Phosphoric Acid, 85%
    Assay ≥85.0% (w/w)
    Grade Certified ACS Grade
       

    DANGER!

    Emergency Overview
    Causes burns by all exposure routes. Use only under a chemical fume hood. Wear personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Do not induce vomiting. Do not induce vomiting. Call a physician or Poison Control Center immediately.

    NFPA

    Health:3
    Flammability:0
    Instability:0

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Product Certifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit