o-Phosphoric Acid (NF/FCC), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.

Packaging Glass Bottle
Quantity 1L

 View more versions of this product

Catalog No. A365-1

  Check Availability
Add to Cart

Custom packing/Bulk ordering

You can now order this item in custom quantities.  Learn More

    Chemical Identifiers

    CAS 7664-38-2, 7732-18-5,7732-18-5
    Molecular Formula H3O4P
    Formula Weight 98
    MDL Number MFCD00011340
    Chemical Name or Material o-Phosphoric Acid
    Assay 85.0 to 88.0% (FCC)
    Grade FCC/NF

    Specifications

    Melting Point 21°C
    Boiling Point 158°C
    Formula Weight 98
    Quantity 1L
    CAS 7664-38-2, 7732-18-5,7732-18-5
    UN Number 1805
    DOT Information DOT Class 8, : Corrosive
    Physical Form Liquid
    Color Clear Viscous
    pH 1
    Alpha Vector Phosphoric Acid
    Density 1.69g/mL
    Vapor Pressure 2 hPa at 20°C
    Health Hazard 3 Emergency Overview
    Causes burns by all exposure routes. Use only under a chemical fume hood. Wear personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Do not induce vomiting. Do not induce vomiting. Call a physician or Poison Control Center immediately.

    NFPA

    Health:3
    Flammability:0
    Instability:0
    Identification Pass Test (FCC)
    Arsenic (As) 3ppm max. (FCC)
    Residual Solvents Meets Requirements (NF)
    Sulfate Pass Test (NF)
    Cleanup Symbol Acid
    MDL Number MFCD00011340
    Alkali Phosphates Pass Test (NF)
    Fluorides 10mg/kg max. (FCC)
    Heavy Metals (as Pb) 0.001% max. (NF)
    Lead (Pb) 3mg/kg max. (FCC)
    Nitrate Pass Test (NF)
    Packaging Glass Bottle
    Phosphorous and Hypophosphorous Acid Pass Test (NF)
    Cadmium (Cd) 3mg/kg max. (FCC)
    Molecular Formula H3O4P
    ChemAlert Storage Symbol White
    Health Hazard 2 DANGER!
    Chemical Name or Material o-Phosphoric Acid
    Assay 85.0 to 88.0% (FCC)
    Grade FCC/NF
       

    DANGER!

    Emergency Overview
    Causes burns by all exposure routes. Use only under a chemical fume hood. Wear personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Do not induce vomiting. Do not induce vomiting. Call a physician or Poison Control Center immediately.

    NFPA

    Health:3
    Flammability:0
    Instability:0

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Product Certifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit