Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Phosphoric Acid (NF/FCC), Fisher Chemical

Packaging Glass Bottle
Quantity 4L

 View more versions of this product

Catalog No. A365-4



  Check Availability
Add to cart

Custom packing/Bulk ordering

You can now order this item in custom quantities. Learn More

Specifications Description & Specifications

Chemical Identifiers

7664-38-2, 7732-18-5,7732-18-5
98
o-Phosphoric Acid
FCC/NF
H3O4P
MFCD00011340
85.0 to 88.0% (NF)

Specifications

Clear Viscous
1
7664-38-2, 7732-18-5,7732-18-5
3mg/kg max. (FCC)
H3O4P
MFCD00011340
1.69g/mL
0.001% max. (NF)
3mg/kg max. (FCC)
Glass Bottle
Meets Requirements (NF)
2 hPa at 20°C
21°C
o-Phosphoric Acid
FCC/NF
4L
Pass Test (NF)
3ppm max. (FCC)
Liquid
98
1805
10mg/kg max. (FCC)
Pass Test (FCC)
Pass Test (NF)
Pass Test (NF)
Pass Test (NF)
Phosphoric Acid
158°C
85.0 to 88.0% (NF)

Safety and Handling

SDS & Safety Documents

DANGER!

Emergency Overview
Causes burns by all exposure routes. Use only under a chemical fume hood. Wear personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Do not induce vomiting. Do not induce vomiting. Call a physician or Poison Control Center immediately.

NFPA

Health:3
Flammability:0
Instability:0

DOTInformation : DOT Class 8, : Corrosive

Alternative Products
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit