Learn More
Phthaldialdehyde, 98%, Thermo Scientific Chemicals
Description
Phthaldialdehyde is used as a fluorescent reagent for checking the assay of amines present in proteins, peptides by capillary electrophoresis and chromatography. It is also used for cholesterol assay in the picomole range as well as for fluorometric determination of histamine, histidine and other amino acids. Poly(phthalaldehyde) is obtained by the polymerization reaction which is used to make a photoresist. The nitrogen by o-phthaldialdehyde assay (NOPA) is a method used in winemaking process to measure the completion of fermentation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phthaldialdehyde is used as a fluorescent reagent for checking the assay of amines present in proteins, peptides by capillary electrophoresis and chromatography. It is also used for cholesterol assay in the picomole range as well as for fluorometric determination of histamine, histidine and other amino acids. Poly(phthalaldehyde) is obtained by the polymerization reaction which is used to make a photoresist. The nitrogen by o-phthaldialdehyde assay (NOPA) is a method used in winemaking process to measure the completion of fermentation.
Solubility
Soluble in acetic acid and methanol. Slightly soluble in water.
Notes
Air, light and moisture sensitive. Store in cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, amines and strong bases.
Specifications
Melting Point | 53°C to 57°C |
Flash Point | 132°C (269°F) |
Assay Percent Range | 98% |
Quantity | 10 g |
UN Number | UN2923 |
Beilstein | 878317 |
Sensitivity | Air Sensitive |
Solubility Information | Soluble in acetic acid and methanol. Slightly soluble in water. |
Formula Weight | 134.14 |
Percent Purity | 98% |
Show More |
Safety and Handling
Hazard Category | H301-H314-H317-H335 |
Hazard Statement | GHS H Statement H300-H314-H318-H317 |
Precautionary Statement | P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S. |
EINECSNumber | 211-402-2 |
RTECSNumber | TH6950000 |
TSCA | Yes |
storageNote1 | Air sensitive |
Recommended Storage | Keep cold; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.