Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Phthaloyl dichloride, 90%, remainder mainly phthalic acid, ACROS Organics™

Click to view available options
Quantity:
5mL
100mL
500mL
Packaging:
Glass bottle
88-95-9
C8H4Cl2O2
203.02
MFCD00000666
FYXKZNLBZKRYSS-UHFFFAOYSA-N
phthaloyl chloride,phthaloyl dichloride,phthalic chloride,phthalyl chloride,phthalic dichloride,phthalic acid dichloride,phthalyl dichloride,1,2-benzenedicarbonyl dichloride,o-phthaloyl dichloride,phthaloyldichloride
6955
benzene-1,2-dicarbonyl chloride
C1=CC=C(C(=C1)C(=O)Cl)C(=O)Cl
88-95-9
C8H4Cl2O2
203.02
MFCD00000666
FYXKZNLBZKRYSS-UHFFFAOYSA-N
phthaloyl chloride,phthaloyl dichloride,phthalic chloride,phthalyl chloride,phthalic dichloride,phthalic acid dichloride,phthalyl dichloride,1,2-benzenedicarbonyl dichloride,o-phthaloyl dichloride,phthaloyldichloride
6955
benzene-1,2-dicarbonyl chloride
C1=CC=C(C(=C1)C(=O)Cl)C(=O)Cl
Specific Gravity 1.4
Quantity 5mL
Refractive Index 1.5674 to 1.5694
Packaging Glass bottle
Infrared Spectrum Authentic
Flash Point 113°C
Color Yellow
Melting Point 12.0°C
Density 1.4000g/mL
Boiling Point 269.0°C to 271.0°C
Assay Percent Range 90%
Linear Formula C6H4(COCl)2
Beilstein 09, 805
Fieser 01,882; 14,263
Merck Index 15, 7487
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in ether, decomposes in alcohol
Formula Weight 203.02
Physical Form Liquid
Percent Purity 90%
Chemical Name or Material o-Phthaloyl dichloride, Remainder mainly phthalic acid
Show More Show Less

GHS P Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger