Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ O-tert-Butyl-L-serine tert-butyl ester hydrochloride, 97%

Click to view available options
Quantity:
5g
25g
51537-21-4
C11H24ClNO3
253.767
MFCD00034850
RDWZQVGVBTYCBD-QRPNPIFTSA-N
h-ser tbu-otbu hcl,h-ser tbu-otbu.hcl,o-tert-butyl-l-serine tert-butyl ester hydrochloride,s-tert-butyl 2-amino-3-tert-butoxy propanoate hydrochloride,o-tert-butyl-l-serine tert-butyl ester hcl,s-tert-butyl 2-amino-3-tert-butoxy propanoate hcl,tert-butyl 2s-2-amino-3-tert-butoxy propanoate hydrochloride,h-ser tbu-otbucl,h-ser but-obut hcl,o-t-butylserine-t-butyl ester hydrochloride
16211506
tert-butyl (2S)-2-amino-3-[(2-methylpropan-2-yl)oxy]propanoate;hydrochloride
CC(C)(C)OCC(C(=O)OC(C)(C)C)N.Cl
51537-21-4
C11H24ClNO3
253.767
MFCD00034850
RDWZQVGVBTYCBD-QRPNPIFTSA-N
h-ser tbu-otbu hcl,h-ser tbu-otbu.hcl,o-tert-butyl-l-serine tert-butyl ester hydrochloride,s-tert-butyl 2-amino-3-tert-butoxy propanoate hydrochloride,o-tert-butyl-l-serine tert-butyl ester hcl,s-tert-butyl 2-amino-3-tert-butoxy propanoate hcl,tert-butyl 2s-2-amino-3-tert-butoxy propanoate hydrochloride,h-ser tbu-otbucl,h-ser but-obut hcl,o-t-butylserine-t-butyl ester hydrochloride
16211506
tert-butyl (2S)-2-amino-3-[(2-methylpropan-2-yl)oxy]propanoate;hydrochloride
CC(C)(C)OCC(C(=O)OC(C)(C)C)N.Cl
Beilstein 4886392
Formula Weight 253.77
Physical Form Crystalline Solid
Percent Purity 97%
Color White
Melting Point 170°C to 177°C
Quantity 25g
Chemical Name or Material O-tert-Butyl-L-serine tert-butyl ester hydrochloride

storageNote1 : 2° to 8°C