Alfa Aesar™ O-tert-Butyl-L-tyrosine, 99%

Manufacturer: Alfa Aesar™ B2257503

Quantity 1g

 View more versions of this product

Catalog No. AAB2257503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 18822-59-8
    Molecular Formula C13H19NO3
    Formula Weight 237.3
    MDL Number MFCD00037202
    Synonym H-Tyr(tBu)-OH
    Chemical Name or Material O-tert-Butyl-L-tyrosine

    Specifications

    Formula Weight 237.3
    Synonym H-Tyr(tBu)-OH
    Molecular Formula C13H19NO3
    MDL Number MFCD00037202
    CAS 18822-59-8
    Melting Point ≈205°C (dec.)
    Quantity 1g
    Chemical Name or Material O-tert-Butyl-L-tyrosine
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit