Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-(Tetrahydropyran-2-yl)hydroxylamine, 96%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
6723-30-4
C5H11NO2
117.15
NLXXVSKHVGDQAT-UHFFFAOYSA-N
o-tetrahydro-2h-pyran-2-yl hydroxylamine,o-tetrahydropyran-2-yl-hydroxylamine,o-oxan-2-yl hydroxylamine,o-tetrahydropyran-2-yl hydroxylamine,2-aminooxy tetrahydro-2h-pyran,o-tetrahydro-pyran-2-yl-hydroxylamine,o-tetrahydropyran-2-ylhydroxylamine,o-tertrahydroxy-pyran-2-yl-hydroxylamine,hydroxylamine, o-tetrahydro-2h-pyran-2-yl,h2nothp
5142091
O-(oxan-2-yl)hydroxylamine
C1CCOC(C1)ON
6723-30-4
C5H11NO2
117.15
NLXXVSKHVGDQAT-UHFFFAOYSA-N
o-tetrahydro-2h-pyran-2-yl hydroxylamine,o-tetrahydropyran-2-yl-hydroxylamine,o-oxan-2-yl hydroxylamine,o-tetrahydropyran-2-yl hydroxylamine,2-aminooxy tetrahydro-2h-pyran,o-tetrahydro-pyran-2-yl-hydroxylamine,o-tetrahydropyran-2-ylhydroxylamine,o-tertrahydroxy-pyran-2-yl-hydroxylamine,hydroxylamine, o-tetrahydro-2h-pyran-2-yl,h2nothp
5142091
O-(oxan-2-yl)hydroxylamine
C1CCOC(C1)ON
Water 1% Max.
Color White
Packaging Glass bottle
Infrared Spectrum Authentic
Melting Point 32.7°C to 37.0°C
Quantity 1g
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 95% min. (GC)
Formula Weight 117.15
Physical Form Low Melting Solid
Percent Purity 96%
Chemical Name or Material O-(Tetrahydropyran-2-yl)hydroxylamine
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning