Alfa Aesar™ o-Thiocresol, 98%

Manufacturer: Alfa Aesar™ B2246022

Quantity 100g

 View more versions of this product

Catalog No. AAB2246022

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 137-06-4
    Molecular Formula C7H8S
    Formula Weight 124.21
    MDL Number MFCD00004838
    UN Number UN2810
    Beilstein 774068
    Merck Index 14,9325
    Synonym 2-Mercaptotoluene; 2-Methylthiophenol
    Chemical Name or Material o-Thiocresol

    Specifications

    Formula Weight 124.21
    TSCA TSCA
    Molecular Formula C7H8S
    MDL Number MFCD00004838
    UN Number UN2810
    Merck Index 14,9325
    Synonym 2-Mercaptotoluene; 2-Methylthiophenol
    Health Hazard 2 GHS H Statement
    H331-H318-H302-H312-H315-H335
    Toxic if inhaled.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 195°C
    Melting Point 10° to 12°C
    Flash Point 63°C (145°F)
    Quantity 100g
    RTECS Number XT8750000
    CAS 137-06-4
    Beilstein 774068
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
    Chemical Name or Material o-Thiocresol
    Assay 98%

    Description

    Applications
    o-Thiocresol is used as a thiol for proteomics research.

    Solubility
    Soluble in ether and ethanol. Insoluble in water.

    Notes
    Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.

    GHS H Statement
    H331-H318-H302-H312-H315-H335
    Toxic if inhaled.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit