Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Tolidine dihydrochloride, ACS reagent, Acros Organics

Click to view available options
Quantity:
25g
100g
500g
612-82-8
C14H16N2·2HCl
285.2
LUKPNZHXJRJBAN-UHFFFAOYSA-N
3,3'-dimethylbenzidine dihydrochloride,o-tolidine dihydrochloride,3,3'-dimethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride,unii-5msk350kd8,dsstox_cid_511,dsstox_rid_75633,dsstox_gsid_20511,tolidine dihydrochloride,1,1'-biphenyl-4,4'-diamine, 3,3'-dimethyl-, dihydrochloride,3,3'-dimethylbenzidine.2hcl
108938
4-(4-amino-3-methylphenyl)-2-methylaniline;dihydrochloride
CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N.Cl.Cl
612-82-8
C14H16N2·2HCl
285.2
LUKPNZHXJRJBAN-UHFFFAOYSA-N
3,3'-dimethylbenzidine dihydrochloride,o-tolidine dihydrochloride,3,3'-dimethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride,unii-5msk350kd8,dsstox_cid_511,dsstox_rid_75633,dsstox_gsid_20511,tolidine dihydrochloride,1,1'-biphenyl-4,4'-diamine, 3,3'-dimethyl-, dihydrochloride,3,3'-dimethylbenzidine.2hcl
108938
4-(4-amino-3-methylphenyl)-2-methylaniline;dihydrochloride
CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N.Cl.Cl
Residue after Ignition 0.1% max
Infrared Spectrum Authentic
Melting Point 130.0°C
Ignition Residue 0.1% max.
Quantity 25g
Merck Index 15,9675
Solubility Information Passes test
Formula Weight 285.2
Grade ACS Reagent
Chemical Name or Material o-Tolidine dihydrochloride

GHS H Statement
May cause cancer.
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Danger

EINECSNumber : 210-322-5