Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ o-Tolidine dihydrochloride, ACS

Click to view available options
Quantity:
25g
100g
500g
612-82-8
C14H18Cl2N2
285.212
MFCD00012960
LUKPNZHXJRJBAN-UHFFFAOYSA-N
3,3'-dimethylbenzidine dihydrochloride,o-tolidine dihydrochloride,3,3'-dimethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride,unii-5msk350kd8,dsstox_cid_511,dsstox_rid_75633,dsstox_gsid_20511,tolidine dihydrochloride,1,1'-biphenyl-4,4'-diamine, 3,3'-dimethyl-, dihydrochloride,3,3'-dimethylbenzidine.2hcl
108938
4-(4-amino-3-methylphenyl)-2-methylaniline;dihydrochloride
CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N.Cl.Cl
612-82-8
C14H18Cl2N2
285.212
MFCD00012960
LUKPNZHXJRJBAN-UHFFFAOYSA-N
3,3'-dimethylbenzidine dihydrochloride,o-tolidine dihydrochloride,3,3'-dimethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride,unii-5msk350kd8,dsstox_cid_511,dsstox_rid_75633,dsstox_gsid_20511,tolidine dihydrochloride,1,1'-biphenyl-4,4'-diamine, 3,3'-dimethyl-, dihydrochloride,3,3'-dimethylbenzidine.2hcl
108938
4-(4-amino-3-methylphenyl)-2-methylaniline;dihydrochloride
CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N.Cl.Cl
Sensitivity Air and light sensitive
Quantity 25g
Odor Odorless
Color White to Cream
Melting Point >300°C
UN Number UN3077
Solubility Information Soluble in water,alcohol,ether and dilute acids
Formula Weight 285.22
Physical Form Solid
Grade ACS
Chemical Name or Material o-Tolidine dihydrochloride
Show More Show Less

GHS H Statement
H350-H302
May cause cancer.
Harmful if swallowed.

P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c

H302-H350

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 210-322-5

RTECSNumber : DD1226000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon