Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Tolidine Dihydrochloride (Certified ACS), Fisher Chemical

Click to view available options
Quantity:
100g
Packaging:
Glass Bottle
612-82-8
C14H18Cl2N2
285.212
MFCD00012960
LUKPNZHXJRJBAN-UHFFFAOYSA-N
3,3'-dimethylbenzidine dihydrochloride,o-tolidine dihydrochloride,3,3'-dimethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride,unii-5msk350kd8,dsstox_cid_511,dsstox_rid_75633,dsstox_gsid_20511,tolidine dihydrochloride,1,1'-biphenyl-4,4'-diamine, 3,3'-dimethyl-, dihydrochloride,3,3'-dimethylbenzidine.2hcl
108938
4-(4-amino-3-methylphenyl)-2-methylaniline;dihydrochloride
CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N.Cl.Cl
612-82-8
C14H18Cl2N2
285.212
MFCD00012960
LUKPNZHXJRJBAN-UHFFFAOYSA-N
3,3'-dimethylbenzidine dihydrochloride,o-tolidine dihydrochloride,3,3'-dimethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride,unii-5msk350kd8,dsstox_cid_511,dsstox_rid_75633,dsstox_gsid_20511,tolidine dihydrochloride,1,1'-biphenyl-4,4'-diamine, 3,3'-dimethyl-, dihydrochloride,3,3'-dimethylbenzidine.2hcl
108938
4-(4-amino-3-methylphenyl)-2-methylaniline;dihydrochloride
CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N.Cl.Cl
Sensitivity Pass Test (0.1ppm free chlorine)
Packaging Glass Bottle
Quantity 100g
Ignition Residue 0.1% max.
Melting Point 130°C
Color Beige
Alpha Vector Tolidine Dihydrochloride
Identification Pass Test
Linear Formula (C6H3-4-NH2-3-CH3)2·2HCl
Formula Weight 285.22
Physical Form Powder/Solid
Grade Certified ACS
Chemical Name or Material o-Tolidine Dihydrochloride
Show More Show Less