Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Tolidine 98.0+%, TCI America™

Click to view available options
Quantity:
25g
100g
500g
119-93-7
C14H16N2
212.296
MFCD00014773
NUIURNJTPRWVAP-UHFFFAOYSA-N
o-tolidine,3,3'-dimethylbenzidine,orthotolidine,diaminoditolyl,2-tolidine,diaminotolyl,bianisidine,3,3'-tolidine,3,3'-dimethylbiphenyl-4,4'-diamine,4,4'-bi-o-toluidine
8413
CHEBI:34320
4-(4-amino-3-methylphenyl)-2-methylaniline
CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N
119-93-7
C14H16N2
212.296
MFCD00014773
NUIURNJTPRWVAP-UHFFFAOYSA-N
o-tolidine,3,3'-dimethylbenzidine,orthotolidine,diaminoditolyl,2-tolidine,diaminotolyl,bianisidine,3,3'-tolidine,3,3'-dimethylbiphenyl-4,4'-diamine,4,4'-bi-o-toluidine
8413
CHEBI:34320
4-(4-amino-3-methylphenyl)-2-methylaniline
CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N
Quantity 100g
Melting Point 130°C
Color Undesignated
Boiling Point 300°C
UN Number 3077
Formula Weight 212.30
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material o-Tolidine

EINECSNumber : (9)-0882

RTECSNumber : DD1225000

TSCA : Yes