Alfa Aesar™ o-Tolualdehyde, 98%, stab. with 0.1% hydroquinone

Manufacturer: Alfa Aesar™ A1017214

Quantity 25g

 View more versions of this product

Catalog No. AAA1017214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 529-20-4
    Molecular Formula C8H8O
    Formula Weight 120.15
    MDL Number MFCD00003338
    Beilstein 605841
    Merck Index 14,9527
    Synonym 2-Methylbenzaldehyde
    Chemical Name or Material o-Tolualdehyde, Stabilized with 0.1% hydroquinone

    Specifications

    Formula Weight 120.15
    Health Hazard 3 GHS P Statement
    P210-P261-P301+P310-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 529-20-4
    Molecular Formula C8H8O
    MDL Number MFCD00003338
    Health Hazard 2 GHS H Statement
    H301-H227-H315-H319-H335
    Toxic if swallowed.
    Combustible liquid.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Beilstein 605841
    Merck Index 14,9527
    Synonym 2-Methylbenzaldehyde
    Health Hazard 1 Danger
    Boiling Point 199° to 200°C
    Flash Point 67°C (152°F)
    Odor Almond-like
    Melting Point −35°C
    Quantity 25g
    Chemical Name or Material o-Tolualdehyde, Stabilized with 0.1% hydroquinone
    Assay 98%

    Description

    Applications
    o-tolualdehyde was used in determination of alkenal-2,4-dinitrophenylhydrazones by HPLC by addition of phosphoric acid. It produces to give a single diastereomer of the tetrahydronaphthalene derivative under UV irradiation.

    Solubility
    Soluble in carbon tetrachloride, ethanol, ethyl ether, benzene. Not miscible in water.

    Notes
    Air Sensitive. Incompatible with air and oxidizing agents. Store under dry inert gas.

    GHS H Statement
    H301-H227-H315-H319-H335
    Toxic if swallowed.
    Combustible liquid.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P301+P310-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit