Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Tolualdehyde, Stabilized 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
250g
Packaging:
Glass bottle
529-20-4,123-31-9
C8H8O
120.15
MFCD00003338
BTFQKIATRPGRBS-UHFFFAOYSA-N
o-tolualdehyde,2-tolualdehyde,2-formyltoluene,o-toluylaldehyde,o-toluic aldehyde,o-tolylaldehyde,o-methylbenzaldehyde,benzaldehyde, 2-methyl,2-methyl-benzaldehyde,o-methylbenazldehyde
10722
CHEBI:27434
2-methylbenzaldehyde
CC1=CC=CC=C1C=O
529-20-4,123-31-9
C8H8O
120.15
MFCD00003338
BTFQKIATRPGRBS-UHFFFAOYSA-N
o-tolualdehyde,2-tolualdehyde,2-formyltoluene,o-toluylaldehyde,o-toluic aldehyde,o-tolylaldehyde,o-methylbenzaldehyde,benzaldehyde, 2-methyl,2-methyl-benzaldehyde,o-methylbenazldehyde
10722
CHEBI:27434
2-methylbenzaldehyde
CC1=CC=CC=C1C=O
Melting Point -35°C
Boiling Point 199°C to 200°C
Flash Point 67°C
Specific Gravity 1.039
Density 1.0390g/mL
Refractive Index 1.5460 to 1.548
Quantity 25g
Packaging Glass bottle
Infrared Spectrum Authentic
Color Colorless to Yellow
Stabilizer 0.1% hydroquinone
Assay Percent Range 98%
Linear Formula CH3C6H4CHO
Beilstein 07, 295
Merck Index 15, 9686
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol and ether
Formula Weight 120.15
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material o-Tolualdehyde, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.
Causes skin irritation.
Toxic in contact with skin.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

AIR SENSITIVE

EINECSNumber : 208-452-2