Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Toluenethiol 97.0+%, TCI America™

Click to view available options
Quantity:
25mL
100mL
500mL
137-06-4
C7H8S
124.201
MFCD00004838
LXUNZSDDXMPKLP-UHFFFAOYSA-N
o-toluenethiol,o-thiocresol,2-methylthiophenol,2-thiocresol,2-toluenethiol,o-tolyl mercaptan,o-mercaptotoluene,o-methylthiophenol,o-methylbenzenethiol,benzenethiol, 2-methyl
8712
2-methylbenzenethiol
CC1=CC=CC=C1S
137-06-4
C7H8S
124.201
MFCD00004838
LXUNZSDDXMPKLP-UHFFFAOYSA-N
o-toluenethiol,o-thiocresol,2-methylthiophenol,2-thiocresol,2-toluenethiol,o-tolyl mercaptan,o-mercaptotoluene,o-methylthiophenol,o-methylbenzenethiol,benzenethiol, 2-methyl
8712
2-methylbenzenethiol
CC1=CC=CC=C1S
Color Yellow
Boiling Point 194°C
Quantity 500mL
Formula Weight 124.20
Physical Form Clear Liquid at 20°C
Percent Purity ≥97.0% (GC)
Chemical Name or Material o-Toluenethiol

RTECSNumber : XT8750000

TSCA : Yes