Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Toluic Acid +98%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
1kg
Packaging:
Glass Bottle
Plastic bottle
118-90-1
C8H8O2
136.15
MFCD00002477
ZWLPBLYKEWSWPD-UHFFFAOYSA-N
o-toluic acid,o-toluylic acid,orthotoluic acid,2-toluic acid,toluic acid,benzoic acid, 2-methyl,o-methylbenzoic acid,2-methyl-benzoic acid,benzoic acid, methyl,o-toluate
8373
CHEBI:36632
2-methylbenzoic acid
CC1=CC=CC=C1C(=O)O
118-90-1
C8H8O2
136.15
MFCD00002477
ZWLPBLYKEWSWPD-UHFFFAOYSA-N
o-toluic acid,o-toluylic acid,orthotoluic acid,2-toluic acid,toluic acid,benzoic acid, 2-methyl,o-methylbenzoic acid,2-methyl-benzoic acid,benzoic acid, methyl,o-toluate
8373
CHEBI:36632
2-methylbenzoic acid
CC1=CC=CC=C1C(=O)O
Boiling Point 258°C to 259°C
Flash Point 148°C
Melting Point 103°C to 105°C
Color White to Yellow
Infrared Spectrum Authentic
Density 1.06
Packaging Glass Bottle
Quantity 5g
Assay Percent Range 98+%
Linear Formula CH3C6H4CO2H
Beilstein 09, 462
Merck Index 14, 9535
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and chloroform
Formula Weight 136.15
Physical Form Needle-Like Crystalline Solid
Percent Purity ≥98.0%
Chemical Name or Material o-Toluic acid, 98+%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 204-284-9

RTECSNumber : XU1400000

TSCA : TSCA