Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ o-Toluidine, 99%

Click to view available options
Quantity:
250g
1000g
5000g
95-53-4
C7H9N
107.156
MFCD00007730
RNVCVTLRINQCPJ-UHFFFAOYSA-N
o-toluidine,2-toluidine,o-tolylamine,2-aminotoluene,2-methylbenzenamine,ortho-toluidine,o-methylaniline,benzenamine, 2-methyl,o-aminotoluene,o-methylbenzenamine
7242
CHEBI:66892
2-methylaniline
CC1=CC=CC=C1N
95-53-4
C7H9N
107.156
MFCD00007730
RNVCVTLRINQCPJ-UHFFFAOYSA-N
o-toluidine,2-toluidine,o-tolylamine,2-aminotoluene,2-methylbenzenamine,ortho-toluidine,o-methylaniline,benzenamine, 2-methyl,o-aminotoluene,o-methylbenzenamine
7242
CHEBI:66892
2-methylaniline
CC1=CC=CC=C1N
Applications
o-Toluidine is used in the production of dyes, as a presumptive test for blood in forensic science. And it is also used in determination of glucose in biological materials, as a precursor in the synthesis of poly(o-toluidine) and copper nanoparticle composite material.

Solubility
Soluble in water(15g/L).

Notes
Air & Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Sensitivity Air and light sensitive
Refractive Index 1.572
Density 1.004
Flash Point 85°C (185°F)
Boiling Point 199°C to 200°C
Odor Unpleasant
Melting Point -24°C
Quantity 250g
UN Number UN1708
Beilstein 741981
Merck Index 14,9536
Solubility Information Soluble in water(15g/L).
Formula Weight 107.16
Percent Purity 99%
Chemical Name or Material o-Toluidine
Show More Show Less

GHS H Statement
H301-H331-H350-H227-H319
Toxic if swallowed.
Toxic if inhaled.
May cause cancer.
Combustible liquid.
Causes serious eye irritation.

P201-P202-P210-P235-P261-P264b-P270-P271-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P311-P312-P330-P332+P313-P362-P370+P378q-P501c

H227-H301+H331-H312-H315-H318-H341-H350

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOLUIDINES, LIQUID

EINECSNumber : 202-429-0

RTECSNumber : XU2975000

TSCA : Yes

Recommended Storage : Ambient temperatures